Learn More
Invitrogen™ SMN1 Recombinant Rabbit Monoclonal Antibody (9N9Q6)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542936
Product Code. 17858092
  • € 524.00 / 100µL
Description
Positive test controls include: HeLa, HepG2. Immunogen sequence: SEDGCIYPAT IASIDFKRET CVVVYTGYGN REEQNLSDLL SPICEVANNI EQNAQENENE SQVSTDESEN SRSPGNKSDN IKPKSAPWNS FLPPPPPM.
The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMN1
Recombinant Monoclonal
1.8 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O35876, Q16637
SMN1
Recombinant fusion protein containing a sequence corresponding to amino acids 103-200 of human SMN1 (Q16637).
100 μL
Primary
Human, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
9N9Q6
Unconjugated
SMN1
AI849087; BCD541; component of gems 1; Gemin1; Gemin-1; SMA; SMA@; SMA1; SMA2; SMA3; SMA4; Smn; SMN1; SMN2; SMNC; SMNT; survival motor neuron 1; survival motor neuron 1 protein; survival motor neuron protein; survival of motor neuron 1, telomeric; survival of motor neuron protein; T-BCD541; TDRD16A; tudor domain containing 16A
Rabbit
Affinity Chromatography
RUO
64301, 6606
-20°C, Avoid Freeze/Thaw Cycles