missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMNDC1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47302H
This item is not returnable.
View return policy
Description
SMNDC1 Polyclonal specifically detects SMNDC1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SMNDC1 | |
| Polyclonal | |
| Western Blot | |
| SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| SMNDC1 | |
| The immunogen for this product maps to a region between residue 188 and 238 of human Splicing Factor 30 using the numbering given in entry NP_005862.1 (GeneID 10285). | |
| 100 μL | |
| Apoptosis | |
| 10285 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction