missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMNDC1 Antibody (PCRP-SMNDC1-1A9), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08232AF647
This item is not returnable.
View return policy
Description
SMNDC1 Monoclonal antibody specifically detects SMNDC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| SMNDC1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30 | |
| Recombinant full-length human SMNDC1 protein (Uniprot: O75940 ) | |
| 100 μg | |
| Apoptosis | |
| 10285 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-SMNDC1-1A9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction