missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SMOC-1 Antibody (09) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26898DL594
This item is not returnable.
View return policy
Description
SMOC-1 Monoclonal antibody specifically detects SMOC-1 in Human samples. It is validated for ELISA
Specifications
| SMOC-1 | |
| Monoclonal | |
| DyLight 594 | |
| Secreted modular calcium-binding protein 1, SMOC-1, SPARC related modular calcium binding 1, SPARC-related modular calcium-binding protein 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SMOC-1 (RefSeq#: NP_001030024.1; Met1-Val435). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 09 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 64093 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction