missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Smooth Surface Filter Discs Qualitative Filter Papers, Grade: 3 w

Used for analytical and technical purposes.

Brand:  Sartorius™ FT-3-308-070

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10659151

  • € 10.10 / Pack of 100
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • For routine analyses like clarification, determination of substances
  • Discs with a center hole for technical applications
  • Made of refined pulp and cotton linters with >95% α-cellulose content
  • Smooth surface
  • Wet-strengthened
Specifications

Specifications

Disc
15μm/sec.
3 w
0.14 mm
70 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers