missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smoothelin Antibody (885424), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8278CL1
This item is not returnable.
View return policy
Description
Smoothelin Monoclonal antibody specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Smoothelin | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ35365, FLJ38597, smoothelin, SMSMO | |
| E. coli-derived recombinant human Smoothelin B, Met1-Leu132, Accession # P53814 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 885424 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6525 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction