missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smoothelin Antibody (885424), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8278K
This item is not returnable.
View return policy
Description
Smoothelin Monoclonal antibody specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Smoothelin | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6525 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 885424 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FLJ35365, FLJ38597, smoothelin, SMSMO | |
| E. coli-derived recombinant human Smoothelin B, Met1-Leu132, Accession # P53814 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction