missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smoothelin Antibody (SMTN/2320R), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20935G
This item is not returnable.
View return policy
Description
Smoothelin Monoclonal antibody specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Smoothelin | |
| Monoclonal | |
| DyLight 488 | |
| FLJ35365, FLJ38597, smoothelin, SMSMO | |
| Recombinant full-length human Smoothelin protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SMTN/2320R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 6525 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction