missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMPD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143767
This item is not returnable.
View return policy
Description
The antibody detects a 78 kDa* band corresponding to nSMase2 on SDS-PAGE immunoblots of adult mouse brain and rat PC12 cells.
Catalyzes the hydrolysis of sphingomyelin to form ceramide and phosphocholine. Ceramide mediates numerous cellular functions, such as apoptosis and growth arrest, and is capable of regulating these 2 cellular events independently. Also hydrolyzes sphingosylphosphocholine. Regulates the cell cycle by acting as a growth suppressor in confluent cells. Probably acts as a regulator of postnatal development and participates in bone and dentin mineralization.
Specifications
| SMPD3 | |
| Polyclonal | |
| Unconjugated | |
| SMPD3 | |
| 4631433G07Rik; AI427456; AW537966; Cca1; confluent 3Y1 cell-associated 1; confluent 3Y1 cell-associated protein 1; fro; neutral sphingomyelin phosphodiesterase 3; neutral sphingomyelinase 2; Neutral sphingomyelinase II; NSMASE2; nSMase-2; SMPD3; Sphingomyelin phosphodiesterase 3; sphingomyelin phosphodiesterase 3, neutral; sphingomyelin phosphodiesterase 3, neutral membrane; sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinase II) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55512, 58994, 94338 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| O35049, Q9JJY3, Q9NY59 | |
| SMPD3 | |
| nSMase2 synthetic peptide corresponds to amino acids in the C-terminal region of human nSMase2. This sequence is highly conserved in rat and mouse nSMase2, and has low homology to other nSMase family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction