missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMPD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143768
This item is not returnable.
View return policy
Description
The antibody detects an 85-90 kDa* band corresponding to nSMase3 on SDS-PAGE immunoblots of adult mouse heart and mouse C2C12 cells. The antibody works for western blot, ELISA, immunoprecipitation, and immunocytochemistry.
The protein encoded by SMPD4 is a sphingomyelinase that catalyzes the hydrolysis of membrane sphingomyelin to form phosphorylcholine and ceramide. This gene is activated by DNA damage, cellular stress, and tumor necrosis factor, but it is downregulated by wild-type p53. The encoded protein localizes to the endoplasmic reticulum and Golgi network.
Specifications
| SMPD4 | |
| Polyclonal | |
| Unconjugated | |
| SMPD4 | |
| 4122402O22Rik; Kiaa1418; mKIAA1418; NET13; neutral membrane (neutral sphingomyelinase-3); neutral sphingomyelinase 3; Neutral sphingomyelinase III; neutral sphingomyelinase-3; NSMASE3; nSMase-3; RGD1310674; SKNY; SKNY protein; Smpd4; Sphingomyelin phosphodiesterase 4; sphingomyelin phosphodiesterase 4, neutral membrane; sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 303790, 55627, 77626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q6ZPR5, Q9NXE4 | |
| SMPD4 | |
| nSMase3 synthetic peptide corresponds to amino acids in the C-terminal region of mouse nSMase3. This sequence is highly conserved in rat and human nSMase3, and has low homology to other nSMase family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction