missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPDL3A Antibody (004), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90229B
This item is not returnable.
View return policy
Description
SMPDL3A Monoclonal specifically detects SMPDL3A in Human samples. It is validated for ELISA.
Specifications
| SMPDL3A | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 4 | |
| ELISA | |
| 0610010C24Rik, acid sphingomyelinase-like phosphodiesterase 3a, ASM3A, ASML3a, ASM-like phosphodiesterase 3a, EC 3.1.4, EC 3.1.4.-, FLJ20177, sphingomyelin phosphodiesterase, acid-like 3A, yR36GH4.1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SMPDL3A (Q92484; Met1-Tyr453). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 10924 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction