Learn More
SMPDL3A Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00152MFV500
Product Code. 18499319
  • € 428.00 / 0.10mL
Description
SMPDL3A Polyclonal antibody specifically detects SMPDL3A in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMPDL3A
mFluor Violet 500 SE
Primary
ELISA
50mM Sodium Borate
Store at 4C in the dark.