missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMPDL3B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15534402
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15534402 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15534402 Supplier Invitrogen™ Supplier No. PA540798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ILWTGDDTPH VPDEKLGEAA VLEIVERLTK LIREVFPDTK VYAALGNHDF Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 85%; Rabbit: 92%; Rat: 85%; Zebrafish: 85%.

Located on chromosome 1, this gene encodes for acid sphingomyelinase-like phosphodiesterase 3b precursor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPDL3B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SMPDL3B
Gene Accession No. Q92485
Gene Alias 1110054A24Rik; Acid sphingomyelinase-like phosphodiesterase 3b; Asml3b; ASM-like phosphodiesterase 3b; ASMLPD; AU045240; im:7137990; si:ch1073-55d10.1; Smpdl3b; sphingomyelin phosphodiesterase acid like 3B; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B
Gene Symbols SMPDL3B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SMPDL3B (aa 88-137).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27293
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.