missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL2 Mouse anti-Human, Clone: OTI2E6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800553
This item is not returnable.
View return policy
Description
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described.Specifications
| OSBPL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H1P3 | |
| OSBPL2 | |
| Human recombit protein fragment corresponding to amino acids 258-480 of human OSBPL2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2E6 | |
| Unconjugated | |
| OSBPL2 | |
| DFNA67, DNFA67, ORP-2, ORP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction