Learn More
SMS Mouse, anti-Human, CoraLite Plus 488, Clone: 1G8E9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68040-20UL
 View more versions of this product
Product Code. 18099825
  • € 140.00 / 20µL
Description
The SMS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMS. This antibody recognizes human antigen. The SMS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
SMS
1G8E9
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
MRSR, SMS, spermine synthase, SPMSY, SpS, SRS
20 μL
6611
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.