missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMS Recombinant Rabbit Monoclonal Antibody (JE61-85)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544855
This item is not returnable.
View return policy
Description
SMS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
| SMS | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P52788, P97355 | |
| SMS | |
| Synthetic peptide within human Spermine synthase aa 100-150/366. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE61-85 | |
| Unconjugated | |
| SMS | |
| AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20603, 363469, 6611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction