missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMURF2 Recombinant Rabbit Monoclonal Antibody (JU32-34)
SMURF2 Recombinant Rabbit Monoclonal Antibody (JU32-34)
Brand: Invitrogen MA545004
This item is not returnable.
View return policy
Description
SMURF2 Recombinant Monoclonal Antibody for Western Blot
Specifications
| SMURF2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9HAU4 | |
| Smurf2 | |
| Recombinant protein within Human SMURF 2 aa 150-340. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| JU32-34 | |
| Unconjugated | |
| Smurf2 | |
| 2810411E22Rik; AI558114; AI649275; E3 ubiquitin ligase SMURF2; E3 ubiquitin-protein ligase SMURF2; HECT-type E3 ubiquitin transferase SMURF2; hSMURF2; RGD1310067; SMAD specific E3 ubiquitin protein ligase 2; SMAD ubiquitination regulatory factor 2; SMAD-specific E3 ubiquitin-protein ligase 2; SMURF2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction