Learn More
SNAI2 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504929S
Encompass_Preferred
 View more versions of this product
Product Code. 17592742
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
SNAI2 Monoclonal antibody specifically detects SNAI2 in Human samples. It is validated for Western Blot
Specifications
Specifications
SNAI2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P20073
Mouse
Affinity chromatography
RUO
310
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1H10
Unconjugated
ANXA7
MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2
Human recombinant protein fragment corresponding to amino acids 105-411 of human ANXA7 produced in E.coli.
30 μL