Learn More
SNAI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12129-1-AP-20UL
 View more versions of this product
Product Code. 16899653
  • € 145.00 / 20µL
Description
The SNAI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNAI2. This antibody recognizes human, mouse antigen. The SNAI2 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
Specifications
SNAI2
Polyclonal
Unconjugated
20 μL
6591
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
333 μg/mL