missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAI3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | SNAI3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689331
|
Novus Biologicals
NBP2-94434-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601012
|
Novus Biologicals
NBP2-94434-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNAI3 Polyclonal antibody specifically detects SNAI3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SNAI3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MGC129606, protein snail homolog 3, SMUC, snail homolog 3 (Drosophila), SNAIL3, Zfp293, zinc finger protein 293, zinc finger protein SNAI3, ZNF293 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 7-186 of human SNAI3 (NP_840101.1). VKTHSSHRVPNYRRLETQREINGACSACGGLVVPLLPRDKEAPSVPGDLPQPWDRSSAVACISLPLLPRIEEALGASGLDALEVSEVDPRASRAAIVPLKDSLNHLNLPPLLVLPTRWSPTLGPDRHGAPEKLLGAERMPRAPGGFECFHCHKPYHTLAGLARHRQLHCHLQVGRVFTCK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 333929 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title