missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Snail Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29626F
This item is not returnable.
View return policy
Description
Snail Polyclonal antibody specifically detects Snail in Human, Mouse, Primate samples. It is validated for Western Blot
Specifications
| Snail | |
| Polyclonal | |
| Western Blot | |
| dJ710H13.1, Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAI1, SNAIL, snail 1 homolog, snail 1 zinc finger protein, snail family zinc finger 1, snail homolog 1, SNAIL1, zinc finger protein SNAI1 | |
| Synthetic peptide made to an internal portion of human SNAIL protein (between residues 200-300) [UniProt O95863] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators | |
| 6615 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction