missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Antibody (4E11), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04344PECY7
This item is not returnable.
View return policy
Description
SNAP25 Monoclonal specifically detects SNAP25 in Human samples. It is validated for Flow Cytometry.
Specifications
| SNAP25 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| SNAP25 | |
| Recombinant human SNAP25 (1-206aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human SNAP25 mAb, clone 4E11, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human SNAP25 protein. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 400000000000 | |
| Flow Cytometry | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| Mouse | |
| Protein G purified | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 6616.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction