missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Antibody (SP12), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-692R
This item is not returnable.
View return policy
Description
SNAP25 Monoclonal antibody specifically detects SNAP25 in Human, Rat, Porcine, Hamster, Gerbil samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SNAP25 | |
| Monoclonal | |
| DyLight 550 | |
| P60880 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Hamster, Gerbil | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SP12 | |
| 50mM Sodium Borate | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| Crude synaptic immunoprecipitate (Human). | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 6616 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction