Learn More
Invitrogen™ NOV Recombinant Rabbit Monoclonal Antibody (HL2726)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556625
Product Code. 30275644
  • € 500.00 / 100µL
Description
NOV Recombinant Monoclonal Antibody for Western Blot, IHC (P), ICC/IF
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NOV
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P48745, Q9QZQ5
CCN3
Recombinant fragment of human CCN3.
100 μL
Primary
Human, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL2726
Unconjugated
CCN3
C130088N23Rik; CCN family member 3; CCN3; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; IGFBP9; IGFBP-9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma overexpressed; nephroblastoma overexpressed gene; nephroblastoma overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NOV; NovH; protein NOV homolog
Rabbit
Protein A
RUO
4856, 81526
-20°C, Avoid Freeze/Thaw Cycles