missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNAP29 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15878843
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15878843 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15878843 Supplier Invitrogen™ Supplier No. PA570437

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 79%; Mouse: 100%; Rabbit: 79%; Rat: 93%; Zebrafish: 82%.

SNAP29 encodes a product that is a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP29
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Snap29
Gene Accession No. Q9Z2P6
Gene Alias 1300018G05Rik; AI891940; AU020222; BB131856; CEDNIK; fc84f09; FLJ21051; golgi SNARE of 32 kDa; Gs32; im:7145101; LOC553460 protein; si:dkey-178e17.4; SNAP29; SNAP-29; Soluble 29 kDa NSF attachment protein; synaptosomal-associated protein 29; synaptosomal-associated protein 29kD; synaptosomal-associated protein, 29kD; synaptosomal-associated protein, 29kDa; synaptosome associated protein 29; synaptosome associated protein 29kDa; vesicle-membrane fusion protein SNAP-29; wu:fc84f09
Gene Symbols Snap29
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat SNAP29.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116500
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.