Learn More
GA International SnapPeel Cryogenic Barcode Labels for Liquid Nitrogen Storage
Product Code. 17820458
Encompass_Preferred
Click to view available options
Diameter (Metric):
25.4 mm
76.2 mm
Unit Size:
Pack of 2000
Pack of 4000
Color:
Blue
Green
Pink
White
Yellow
Product Code. 17820458
Brand: GA International JTTA560SBC12BL
Only null left
Add to basket
Add to basket
Thermal-transfer cryogenic labels for cryo vials, microtubes, cryo/freezer boxes, etc. destined for storage in liquid and vapor phase liquid nitrogen, laboratory freezers, or transport on dry ice.
  • Resistant to long-term cryogenic storage in freezers and liquid nitrogen, up to -196°C/-321°F
  • Perfect for cryovials, tubes, cryo-boxes, freezer-boxes, and many other cryo-containers
  • Resistant to spraying and swabbing with alcohol and surface sanitizing wipes
  • Require no cure time before liquid nitrogen immersion. Will also adhere to wet surfaces
  • Ideal for serialized data and barcodes, thanks to high-quality thermal-transfer printing
  • Each row has one rectangular and one round label
  • Available in a variety of colors and sizes
  • Stored on a 25.4 mm roll
Specifications
Adhesive Type Permanent
Finish Matte
Format Roll
Material Thermoplastic Film
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Labels per Row 2 Labels
Quantity 2000/Pk.
Shape Rectangle with Circle
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Dimensions (LxW) 23.9mm x 12.7mm + 11.1 mm
Length (English) 0.5 in.
Length (Metric) 12.7 mm
Width (English) 0.94 in.
Width (Metric) 23.9 mm
Diameter (English) 1 in.
Diameter (Metric) 25.4 mm