missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNCB Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68046-1-IG-20UL
This item is not returnable.
View return policy
Description
The SNCB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNCB. This antibody recognizes human, mouse, rat, pig, rabbit, chicken antigen. The SNCB antibody has been validated for the following applications: WB, IF-P, ELISA analysis.Specifications
| SNCB | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6620 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, Immunofluorescence, ELISA | |
| 2B10E10 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit, Chicken | |
| 18 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction