missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SND1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10760-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against near the C-terminus of human SND1.
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, it functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activator of EBNA-2. It also interacts with EAV, NSP1,GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.Specifications
| SND1 | |
| Polyclonal | |
| Unconjugated | |
| SND1 | |
| 100 kDa coactivator, EBNA2 coactivator p100, p100, p100 co activator, SND1, TDRD11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27044, 56463, 64635 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q66X93, Q78PY7, Q7KZF4 | |
| Snd1 | |
| SND1 Fusion Protein Ag1200 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction