missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552676
This item is not returnable.
View return policy
Description
Immunogen sequence: NDLEKSVEKI QRDVSHNHRL VPGPELEEKA LVLKQLGETL TELKAHFPGL QSKMRVVLRV EVEAVKFLKE EPQRLDGLLK RCRGVTDTLA QIRR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
Specifications
| SNIP | |
| Polyclonal | |
| Unconjugated | |
| SRCIN1 | |
| KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0H9 | |
| SRCIN1 | |
| Recombinant protein corresponding to Human SNIP. Recombinant protein control fragment (Product #RP-90631). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction