missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNIP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98468AF532
This item is not returnable.
View return policy
Description
SNIP1 Polyclonal specifically detects SNIP1 in Human samples. It is validated for Western Blot.
Specifications
| SNIP1 | |
| Polyclonal | |
| Western Blot | |
| dJ423B22.2, FHA domain-containing protein SNIP1, FLJ12553, RP3-423B22.3, Smad nuclear interacting protein (SNIP1), Smad nuclear interacting protein 1, smad nuclear-interacting protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human SNIP1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 79753 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction