Learn More
Invitrogen™ SNRNP200 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115942
Product Code. 17248723
  • € 496.00 / 100µL
Description
Antibody detects endogenous levels of total SNRNP200.
Pre-mRNA splicing is catalyzed by the spliceosome, a complex of specialized RNA and protein subunits that removes introns from a transcribed pre-mRNA segment. The spliceosome consists of small nuclear RNA proteins (snRNPs) U1, U2, U4, U5 and U6, together with approximately 80 conserved proteins. U5 snRNP contains nine specific proteins. This gene encodes one of the U5 snRNP-specific proteins. This protein belongs to the DEXH-box family of putative RNA helicases. It is a core component of U4/U6-U5 snRNPs and appears to catalyze an ATP-dependent unwinding of U4/U6 RNA duplices. Mutations in this gene cause autosomal-dominant retinitis pigmentosa. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNRNP200
Polyclonal
Unconjugated
SNRNP200
A330064G03Rik; Activating signal cointegrator 1 complex subunit 3-like 1; ASCC3L1; bad response to refrigeration 2 homolog; BC011390; BRR2; BRR2 homolog; HELIC2; Kiaa0788; RP33; small nuclear ribonucleoprotein 200 (U5); small nuclear ribonucleoprotein 200kDa (U5); small nuclear ribonucleoprotein U5 subunit 200; small nuclear ribonucleoprotein, U5 200kDa subunit; SNRNP200; U5 small nuclear ribonucleoprotein 200 kDa helicase; U5 snRNP-specific 200 kDa protein; U5 snRNP-specific protein, 200 kDa (DEXH RNA helicase family); U5-200KD; U5-200-KD
Rabbit
Affinity chromatography
RUO
23020, 296126, 320632
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
F1LNJ2, O75643, Q6P4T2
SNRNP200
A synthesized peptide derived from human SNRNP200(Accession O75643), corresponding to amino acid residues E1854-L1904.