missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPA1 Recombinant Rabbit Monoclonal Antibody (JE56-64)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544901
This item is not returnable.
View return policy
Description
SNRPA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
SNRPA1 (U2 snRNP-A) is located in gene map locus 15q26. 3 and is a 28 kDa protein, isa specific component of U2 snRNP particle and consists of a unique N-terminal leucine-rich region and an extremely hydrophobic RNA-binding C-terminal region. It lacks segments homologous to RNP1 and RNP2, commonA. A motifs found in several RNA-binding proteins. It associates with U2 snRNP and participates in transcription and pre-mRNA splicing. It might play an important role in cell division and proliferation. Patients with connective tissue disorders often produce autoantibodies against snRNP particles.
Specifications
| SNRPA1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P09661, P57784 | |
| SNRPA1 | |
| Recombinant protein within human SNRPA1 aa 155-255. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE56-64 | |
| Unconjugated | |
| SNRPA1 | |
| 1500015N06Rik; Lea1; small nuclear ribonucleoprotein polypeptide A'; Snrpa1; U2 small nuclear ribonucleoprotein A'; U2 small nuclear ribonucleoprotein polypeptide A'; U2 snRNP A'; U2 snRNP-A' | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100361269, 6627, 68981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction