missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPB2 Antibody (OTI2A5), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74260AF594
This item is not returnable.
View return policy
Description
SNRPB2 Monoclonal specifically detects SNRPB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SNRPB2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SNRPB2 | |
| Full length recombinant protein of human SNRPB2 (NP_937863) produced in E.coli. | |
| Immunology | |
| 6629 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2A5 | |
| Western Blot, Immunohistochemistry | |
| MGC24807, MGC45309, Msl1, small nuclear ribonucleoprotein polypeptide B, small nuclear ribonucleoprotein polypeptide B′′, small nuclear ribonucleoprotein polypeptide B2, U2 small nuclear ribonucleoprotein B′′, U2 snRNP B′′ | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction