missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106378
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SNRPB2.
SNRPB2 is involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A' protein.
Specifications
| SNRPB2 | |
| Polyclonal | |
| Unconjugated | |
| SNRPB2 | |
| 2810052G09Rik; Msl1; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptide B''; small nuclear ribonucleoprotein polypeptide B2; Snrpb2; U2 small nuclear ribonucleoprotein B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''; U2B'' | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20639, 362223, 6629 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08579, Q9CQI7 | |
| SNRPB2 | |
| A synthesized peptide derived from human SNRPB2(Accession P08579), corresponding to amino acid residues I40-I90. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction