missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38755
This item is not returnable.
View return policy
Description
SNRPC Polyclonal specifically detects SNRPC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SNRPC | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P09234 | |
| SNRPC | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VKDYYQKWMEEQAQSLIDKTTAAFQQGKIPPTPFSAPPPAGAMIPPP | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6631 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ20302, small nuclear ribonucleoprotein polypeptide C, U1 small nuclear ribonucleoprotein C, U1 small nuclear RNP specific C, U1 snRNP C, U1 snRNP protein C, U1C, U1-C, Yhc1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction