missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103307
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SmD3.
This gene encodes a core component of the spliceosome, which is a nuclear ribonucleoprotein complex that functions in pre-mRNA splicing. Alternative splicing results in multiple transcript variants.
Specifications
| SNRPD3 | |
| Polyclonal | |
| Unconjugated | |
| Snrpd3 | |
| 1700043E15Rik; 2310009E13Rik; AW046420; small nuclear ribonucleoprotein D3; small nuclear ribonucleoprotein D3 polypeptide; small nuclear ribonucleoprotein D3 polypeptide 18kDa; small nuclear ribonucleoprotein Sm D3; SMD3; Sm-D3; snRNP core protein D3; SNRPD3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6634, 67332, 687711 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62318, P62320 | |
| Snrpd3 | |
| A synthesized peptide derived from human SNRPD3(Accession P62318), corresponding to amino acid residues L58-A108. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction