Learn More
SNS-032, MedChemExpress
Product Code. 16492768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492768 10 mg 10mg
16485507 5 mg 5mg
16480896 200 mg 200mg
16474226 100 mg 100mg
16469466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16492768 Supplier Medchem Express Supplier No. HY10008
MedChemExpress SNS-032 (BMS-387032) is a potent and selective inhibitor of CDK2, CDK7, and CDK9 with IC50s of 38 nM, 62 nM and 4 nM, respectively. SNS-032 has antitumor effect.
SNS-032 (BMS-387032) is a potent and selective inhibitor of CDK2, CDK7, and CDK9 with IC50s of 38 nM, 62 nM and 4 nM, respectively. SNS-032 has antitumor effect.
Specifications
Chemical Name or Material SNS-032
CAS 345627-80-7
Color White
Purity Grade Notes Research
Molecular Formula C17H24N4O2S2
Quantity 10 mg
Synonym BMS-387032
Solubility Information DMSO : 100 mg/mL (262.79 mM; Need ultrasonic)
SMILES CC(C)(C1=CN=C(CSC2=CN=C(NC(C3CCNCC3)=O)S2)O1)C
Molecular Weight (g/mol) 380.53
Formula Weight 380.53
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only