Learn More
SNTA1 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502041
Encompass_Preferred
 View more versions of this product
Product Code. 17545821
  • € 540.87 / 100µL
Description
Syntrophin alpha-1 (Dystrophin, SNTA1) is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
Specifications
Specifications
SNTA1
Monoclonal
0.28 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13424
SNTA1
Full length human recombit protein of human SNTA1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2D5
Unconjugated
SNTA1
LQT12, SNT1, TACIP1, dJ1187J4.5
Mouse
Affinity Chromatography
RUO
6640
-20° C, Avoid Freeze/Thaw Cycles