missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated MARCKS Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-20661-100UL
This item is not returnable.
View return policy
Description
The MARCKS antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MARCKS. This antibody recognizes human, mouse, rat antigen. The MARCKS antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| MARCKS | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4082 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| 80K-L; FLJ14368; FLJ90045; MACS; PKCSL; PRKCSL | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 80 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction