missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNTG1 Antibody (OTI3G10), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74261FR
This item is not returnable.
View return policy
Description
SNTG1 Monoclonal specifically detects SNTG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SNTG1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| SNTG1 | |
| Full length recombinant protein of human SNTG1 (NP_061840) produced in HEK293T cells. | |
| 0.1 mL | |
| 54212.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3G10 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| G1SYNsyntrophin-4, gamma-1-syntrophin, gamma1-syntrophin, SYN4syntrophin 4, syntrophin, gamma 1, Syntrophin-4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction