Learn More
Invitrogen™ SNTG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140012
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ILRFYTAQDG TDWLRAVSAN IRELTLQNMK MANKCCSPSD QVVHMGWVNE KLQGADSSQT FRPKFLALK Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 77%.
Syntrophin gamma 2 encodes a protein belonging to the syntrophin family. Syntrophins are cytoplasmic peripheral membrane proteins that bind to components of mechanosenstive sodium channels and the extreme carboxy-terminal domain of dystrophin and dystrophin-related proteins. The PDZ domain of this protein product interacts with a protein component of a mechanosensitive sodium channel that affects channel gating. Absence or reduction of this protein product has been associated with Duchenne muscular dystrophy. There is evidence of alternative splicing yet the full-length nature of these variants has not been described.
Specifications
| SNTG2 | |
| Polyclonal | |
| Unconjugated | |
| SNTG2 | |
| 2210008K22Rik; 9530013L23; BB121248; G2SYN; Gamma-2-syntrophin; gamma2-syntrophin; MGC133174; SNTG2; SYN5; syntrophin; syntrophin 5; syntrophin gamma 2; syntrophin, gamma 2; syntrophin-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY99 | |
| SNTG2 | |
| Recombinant protein fragment of Human SNTG2 (Product #RP-109083) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.