missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNUPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119423
This item is not returnable.
View return policy
Description
SNUPN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications
| SNUPN | |
| Polyclonal | |
| Unconjugated | |
| SNUPN | |
| 0610031A09Rik; hypothetical protein LOC515588; KPNBL; RNA U transporter 1; RNA, U transporter 1; Rnut1; Snupn; Snupn1; snurportin 1; Snurportin1; snurportin-1; snurportin-1-like protein; SPN1; wu:fc41a03; zgc:77819 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 66069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q80W37 | |
| SNUPN | |
| Recombinant protein Snurportin 1. The antigen corresponds to amino acid range 1-329 of the target protein. | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction