missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNUPN Polyclonal Antibody

Product Code. 17782984
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17782984 20 μL 20µL
17790824 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17782984 Supplier Invitrogen™ Supplier No. PA5121590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNUPN Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNUPN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SNUPN
Gene Accession No. O95149, Q80W37
Gene Alias 0610031A09Rik; hypothetical protein LOC515588; KPNBL; RNA U transporter 1; RNA, U transporter 1; Rnut1; Snupn; Snupn1; snurportin 1; Snurportin1; snurportin-1; snurportin-1-like protein; SPN1; wu:fc41a03; zgc:77819
Gene Symbols SNUPN
Host Species Rabbit
Immunogen Recombinant protein Snurportin 1. The antigen corresponds to amino acid range 1-360 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10073, 66069
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.