missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNURPORTIN1 Antibody (SNUPN/7351R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20883AF700
This item is not returnable.
View return policy
Description
SNURPORTIN1 Monoclonal antibody specifically detects SNURPORTIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SNURPORTIN1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| KPNBL, RNA U transporter 1, RNA, U transporter 1, RNUT1, snurportin 1, Snurportin1, SNURPORTIN-1, SPN1 | |
| Recombinant fragment (around aa 900-1100) of human SNURPORTIN1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SNUPN/7351R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 10073 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction