missing translation for 'onlineSavingsMsg'
Learn More

SNURPORTIN1 Antibody (SNUPN/7351R), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20883F

Product Code. 18661925

  • € 726.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SNURPORTIN1 Monoclonal antibody specifically detects SNURPORTIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNURPORTIN1
Monoclonal
FITC
KPNBL, RNA U transporter 1, RNA, U transporter 1, RNUT1, snurportin 1, Snurportin1, SNURPORTIN-1, SPN1
Recombinant fragment (around aa 900-1100) of human SNURPORTIN1 protein (exact sequence is proprietary)
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
SNUPN/7351R
PBS
Rabbit
Protein A or G purified
RUO
10073
Store at 4°C in the dark.
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.