missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SNW1 Antibody (PCRP-SNW1-1C12) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26930R
This item is not returnable.
View return policy
Description
SNW1 Monoclonal antibody specifically detects SNW1 in Human samples. It is validated for Flow Cytometry
Specifications
| SNW1 | |
| Monoclonal | |
| DyLight 550 | |
| Bx42, homolog of Drosophila BX42, MGC119379, NCOA-62, Nuclear protein SkiP, Nuclear receptor coactivator NCoA-62, nuclear receptor coactivator, 62-kD, Prp45, PRPF45, SKI interacting protein, Ski-interacting protein, SKIPSKIIP, SNW domain containing 1, SNW domain-containing protein 1 | |
| Recombinant fragment of human SNW1 protein | |
| 0.1 mL | |
| Signal Transduction | |
| 22938 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| PCRP-SNW1-1C12 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur