missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119879
This item is not returnable.
View return policy
Description
Positive Control: PC-12 cell lysates, NIH/3T3 cell lysates, Jurkat cell lysates, human kidney tissue, mouse colon tissue, rat testis tissue, Hela, NIH/3T3. Subcellular Location: Nucleus.
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Specifications
| SNW1 | |
| Polyclonal | |
| Unconjugated | |
| Snw1 | |
| 2310008B08Rik; AW048543; Bx42; homolog of Drosophila BX42; NCOA-62; Nuclear protein SkiP; nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; PPS; Prp45; PRPF45; RGD1561926; SKI interacting protein; ski-interacting protein; Skiip; SKIP; SNW domain containing 1; SNW domain-containing protein 1; SNW1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22938, 500695, 66354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q13573, Q9CSN1 | |
| Snw1 | |
| Recombinant protein within human SNW1 aa 1-200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction