missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582586
This item is not returnable.
View return policy
Description
Immunogen sequence: MSNALAIQVD SEGKIKYDAI ARQGQSKDKV IYSKYTDLVP KEVMNADDPD LQRPDEEAIK EITEKTRVAL EKSVSQKVAA AMPVRAADKL APAQYIRYTP SQQGVAFNSG AKQRVIRMVE MQKDPMEPPR FKINKK Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Specifications
| SNW1 | |
| Polyclonal | |
| Unconjugated | |
| Snw1 | |
| 2310008B08Rik; AW048543; Bx42; homolog of Drosophila BX42; NCOA-62; Nuclear protein SkiP; nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; PPS; Prp45; PRPF45; RGD1561926; SKI interacting protein; ski-interacting protein; Skiip; SKIP; SNW domain containing 1; SNW domain-containing protein 1; SNW1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22938, 500695, 66354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13573, Q9CSN1 | |
| Snw1 | |
| Recombinant protein corresponding to Human SNW1. Recombinant protein control fragment (Product #RP-88944). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction