missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118274
This item is not returnable.
View return policy
Description
SNW1 Polyclonal Antibody for Western Blot, IP
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Specifications
| SNW1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q13573 | |
| Snw1 | |
| E. coli-derived Human SNW1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Snw1 | |
| 2310008B08Rik; AW048543; Bx42; homolog of Drosophila BX42; NCOA-62; Nuclear protein SkiP; nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; PPS; Prp45; PRPF45; RGD1561926; SKI interacting protein; ski-interacting protein; Skiip; SKIP; SNW domain containing 1; SNW domain-containing protein 1; SNW1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 22938 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction