missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNW1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25926-1-AP-150UL
This item is not returnable.
View return policy
Description
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.Specifications
| SNW1 | |
| Polyclonal | |
| Unconjugated | |
| SNW1 | |
| Bx42, NCOA 62, Nuclear protein SkiP, Prp45, PRPF45, Ski interacting protein, SKIIP, SKIP, SNW domain containing 1, SNW1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22938 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13573 | |
| SNW1 | |
| SNW1 Fusion Protein Ag23106 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction